Assay Details
| Gene Symbol: LOC4337093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780492.2 | XP_015635978.1 | 12 - 13 | 2309 | 77 | |
| XM_015780493.2 | XP_015635979.1 | 13 - 14 | 2234 | 77 | ||
| GenBank mRNA | AK061702.1 | - | 3 - 4 | 456 | 77 | |
| AK064973.1 | - | 12 - 13 | 1969 | 77 | ||
| AK119202.1 | - | 3 - 4 | 456 | 77 | ||
| AK122184.1 | - | 13 - 14 | 2179 | 77 | ||
More Information
Set Membership: |
|

LOC4337093
Genomic Map