Assay Details
| Gene Symbol: LOC4337093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780492.2 | XP_015635978.1 | 14 - 15 | 2496 | 91 | |
| XM_015780493.2 | XP_015635979.1 | 15 - 16 | 2421 | 91 | ||
| GenBank mRNA | AK061702.1 | - | 5 - 6 | 643 | 91 | |
| AK064973.1 | - | 14 - 15 | 2156 | 91 | ||
| AK119202.1 | - | 5 - 6 | 643 | 91 | ||
| AK122184.1 | - | 15 - 16 | 2366 | 91 | ||
More Information
Set Membership: |
|

LOC4337093
Genomic Map