Assay Details
| Gene Symbol: LOC4337093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780492.2 | XP_015635978.1 | 15 - 16 | 2600 | 92 | |
| XM_015780493.2 | XP_015635979.1 | 16 - 17 | 2525 | 92 | ||
| GenBank mRNA | AK061702.1 | - | 6 - 7 | 747 | 92 | |
| AK064973.1 | - | 15 - 16 | 2260 | 92 | ||
| AK119202.1 | - | 6 - 7 | 747 | 92 | ||
| AK122184.1 | - | 16 - 17 | 2470 | 92 | ||
More Information
Set Membership: |
|

LOC4337093
Genomic Map