Assay Details
| Gene Symbol: LOC4337093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780492.2 | XP_015635978.1 | 6 - 7 | 1187 | 149 | |
| XM_015780493.2 | XP_015635979.1 | 6 - 7 | 1187 | 149 | ||
| GenBank mRNA | AK064973.1 | - | 6 - 7 | 847 | 149 | |
| AK122184.1 | - | 6 - 7 | 1132 | 149 | ||
More Information
Set Membership: |
|

LOC4337093
Genomic Map