Assay Details
| Gene Symbol: LOC4337211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777876.1 | XP_015633362.1 | 4 - 5 | 720 | 66 | |
| GenBank mRNA | AK064227.1 | - | 4 - 5 | 708 | 66 | |
| AK071891.1 | - | 4 - 5 | 728 | 66 | ||
More Information
Set Membership: |
|

LOC4337211
Genomic Map