Assay Details
| Gene Symbol: LOC4337211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777875.2 | XP_015633361.1 | 7 - 8 | 970 | 130 | |
| XM_015777876.1 | XP_015633362.1 | 6 - 7 | 922 | 130 | ||
| GenBank mRNA | AK071891.1 | - | 6 - 7 | 930 | 130 | |
More Information
Set Membership: |
|

LOC4337211
Genomic Map