Assay Details
| Gene Symbol: LOC4337211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777875.2 | XP_015633361.1 | 9 - 10 | 1282 | 90 | |
| XM_015777876.1 | XP_015633362.1 | 8 - 9 | 1234 | 90 | ||
| GenBank mRNA | AK064227.1 | - | 8 - 9 | 1222 | 90 | |
| AK071891.1 | - | 8 - 9 | 1242 | 90 | ||
More Information
Set Membership: |
|

LOC4337211
Genomic Map