Assay Details
| Gene Symbol: LOC4337257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779939.1 | XP_015635425.1 | 10 - 11 | 2354 | 73 | |
| GenBank mRNA | AK073066.1 | - | 7 - 8 | 1284 | 73 | |
| AK100435.1 | - | 10 - 11 | 2355 | 73 | ||
| AK105082.1 | - | 7 - 8 | 910 | 73 | ||
| AY224445.1 | - | 6 - 7 | 705 | 73 | ||
More Information
Set Membership: |
|

LOC4337257
Genomic Map