Assay Details
| Gene Symbol: LOC4337257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779939.1 | XP_015635425.1 | 11 - 12 | 2522 | 89 | |
| GenBank mRNA | AK073066.1 | - | 8 - 9 | 1452 | 89 | |
| AK100435.1 | - | 11 - 12 | 2523 | 89 | ||
| AK105082.1 | - | 8 - 9 | 1078 | 89 | ||
| AY224445.1 | - | 7 - 8 | 873 | 89 | ||
More Information
Set Membership: |
|

LOC4337257
Genomic Map