Assay Details
| Gene Symbol: LOC4337257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779939.1 | XP_015635425.1 | 13 - 14 | 2875 | 59 | |
| GenBank mRNA | AK073066.1 | - | 10 - 11 | 1805 | 59 | |
| AK100435.1 | - | 13 - 14 | 2876 | 59 | ||
| AK105082.1 | - | 10 - 11 | 1431 | 59 | ||
| AY224445.1 | - | 9 - 10 | 1226 | 59 | ||
More Information
Set Membership: |
|

LOC4337257
Genomic Map