Assay Details
| Gene Symbol: LOC4337257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779939.1 | XP_015635425.1 | 14 - 15 | 3068 | 116 | |
| GenBank mRNA | AK073066.1 | - | 11 - 12 | 1998 | 116 | |
| AK100435.1 | - | 14 - 15 | 3069 | 116 | ||
| AK105082.1 | - | 11 - 12 | 1624 | 116 | ||
| AY224445.1 | - | 10 - 11 | 1419 | 116 | ||
| BT131596.1 | - | 1 - 2 | 161 | 116 | ||
More Information
Set Membership: |
|

LOC4337257
Genomic Map