Assay Details
| Gene Symbol: LOC4337257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779939.1 | XP_015635425.1 | 3 - 4 | 707 | 85 | |
| GenBank mRNA | AK100435.1 | - | 3 - 4 | 708 | 85 | |
More Information
Set Membership: |
|

LOC4337257
Genomic Map