Assay Details
| Gene Symbol: LOC4337257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779939.1 | XP_015635425.1 | 4 - 5 | 1588 | 130 | |
| GenBank mRNA | AK073066.1 | - | 1 - 2 | 518 | 130 | |
| AK100435.1 | - | 4 - 5 | 1589 | 130 | ||
| AK105082.1 | - | 1 - 2 | 144 | 130 | ||
More Information
Set Membership: |
|

LOC4337257
Genomic Map