Assay Details
| Gene Symbol: LOC4337257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779939.1 | XP_015635425.1 | 5 - 6 | 1693 | 61 | |
| GenBank mRNA | AK073066.1 | - | 2 - 3 | 623 | 61 | |
| AK100435.1 | - | 5 - 6 | 1694 | 61 | ||
| AK105082.1 | - | 2 - 3 | 249 | 61 | ||
| AY224445.1 | - | 1 - 2 | 44 | 61 | ||
More Information
Set Membership: |
|

LOC4337257
Genomic Map