Assay Details
| Gene Symbol: LOC4337352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777941.2 | XP_015633427.1 | 2 - 3 | 306 | 56 | |
| XM_015777945.2 | XP_015633431.1 | 2 - 3 | 305 | 56 | ||
| GenBank mRNA | AK103099.1 | - | 2 - 3 | 305 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777941.2, XM_015777945.2 transcripts.
Set Membership: |
|

LOC4337352
Genomic Map