Assay Details
| Gene Symbol: LOC4337352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777937.2 | XP_015633423.1 | 7 - 8 | 836 | 119 | |
| XM_015777938.2 | XP_015633424.1 | 7 - 8 | 836 | 119 | ||
| XM_015777939.2 | XP_015633425.1 | 7 - 8 | 836 | 119 | ||
| XM_015777941.2 | XP_015633427.1 | 7 - 8 | 836 | 119 | ||
| XM_015777942.2 | XP_015633428.1 | 7 - 8 | 752 | 119 | ||
| XM_015777943.2 | XP_015633429.1 | 7 - 8 | 656 | 119 | ||
| XM_015777944.2 | XP_015633430.1 | 7 - 8 | 789 | 119 | ||
| XM_015777945.2 | XP_015633431.1 | 7 - 8 | 835 | 119 | ||
| GenBank mRNA | AK103099.1 | - | 7 - 8 | 835 | 119 | |
More Information
Set Membership: |
|

LOC4337352
Genomic Map