Assay Details
| Gene Symbol: LOC4337352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777938.2 | XP_015633424.1 | 8 - 9 | 950 | 79 | |
| XM_015777945.2 | XP_015633431.1 | 8 - 9 | 949 | 79 | ||
More Information
Set Membership: |
|

LOC4337352
Genomic Map