Assay Details
| Gene Symbol: LOC4337461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781693.2 | XP_015637179.1 | 1 | 950 | 81 | |
| GenBank mRNA | AK070446.1 | - | 1 | 954 | 81 | |
| AK104363.1 | - | 1 | 951 | 81 | ||
More Information
Set Membership: |
|

LOC4337461
Genomic Map