Assay Details
| Gene Symbol: LOC4337462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781692.2 | XP_015637178.1 | 1 | 1308 | 120 | |
| GenBank mRNA | AK061257.1 | - | 1 | 877 | 120 | |
| AK069353.1 | - | 1 | 874 | 120 | ||
| AK103610.1 | - | 1 | 1251 | 120 | ||
More Information
Set Membership: |
|

LOC4337462
Genomic Map