Assay Details
| Gene Symbol: LOC4337579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785137.2 | XP_015640623.1 | 5 - 6 | 918 | 108 | |
| GenBank mRNA | AK070061.1 | - | 5 - 6 | 924 | 108 | |
| AY336987.1 | - | 4 - 5 | 802 | 108 | ||
More Information
Set Membership: |
|

LOC4337579
Genomic Map