Assay Details
| Gene Symbol: LOC4337579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785137.2 | XP_015640623.1 | 6 - 7 | 1099 | 143 | |
| GenBank mRNA | AK070061.1 | - | 6 - 7 | 1105 | 143 | |
More Information
Set Membership: |
|

LOC4337579
Genomic Map