Assay Details
| Gene Symbol: LOC4337963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782393.2 | XP_015637879.1 | 10 - 11 | 1431 | 125 | |
| XM_015782394.2 | XP_015637880.1 | 10 - 11 | 1342 | 125 | ||
| XM_015782395.2 | XP_015637881.1 | 10 - 11 | 1422 | 125 | ||
| GenBank mRNA | DQ421409.1 | - | 9 - 10 | 1244 | 125 | |
More Information
Set Membership: |
|

LOC4337963
Genomic Map