Assay Details
| Gene Symbol: LOC4337963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782393.2 | XP_015637879.1 | 4 - 5 | 567 | 95 | |
| XM_015782394.2 | XP_015637880.1 | 4 - 5 | 478 | 95 | ||
| XM_015782395.2 | XP_015637881.1 | 4 - 5 | 558 | 95 | ||
| GenBank mRNA | DQ421409.1 | - | 3 - 4 | 380 | 95 | |
More Information
Set Membership: |
|

LOC4337963
Genomic Map