Assay Details
| Gene Symbol: LOC4337963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782393.2 | XP_015637879.1 | 5 - 6 | 706 | 94 | |
| XM_015782394.2 | XP_015637880.1 | 5 - 6 | 617 | 94 | ||
| XM_015782395.2 | XP_015637881.1 | 5 - 6 | 697 | 94 | ||
| GenBank mRNA | DQ421409.1 | - | 4 - 5 | 519 | 94 | |
More Information
Set Membership: |
|

LOC4337963
Genomic Map