Assay Details
| Gene Symbol: LOC4338081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783795.2 | XP_015639281.1 | 10 - 11 | 1319 | 113 | |
| XM_015783796.1 | XP_015639282.1 | 10 - 11 | 1098 | 113 | ||
| XM_015783797.2 | XP_015639283.1 | 10 - 11 | 1140 | 113 | ||
| GenBank mRNA | AK068570.1 | - | 7 - 8 | 892 | 113 | |
| AK068965.1 | - | 11 - 12 | 1316 | 113 | ||
More Information
Set Membership: |
|

LOC4338081
Genomic Map