Assay Details
| Gene Symbol: LOC4338081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783795.2 | XP_015639281.1 | 12 - 13 | 1470 | 136 | |
| XM_015783796.1 | XP_015639282.1 | 12 - 13 | 1249 | 136 | ||
| XM_015783797.2 | XP_015639283.1 | 12 - 13 | 1291 | 136 | ||
| GenBank mRNA | AK068570.1 | - | 9 - 10 | 1043 | 136 | |
| AK068965.1 | - | 13 - 14 | 1467 | 136 | ||
More Information
Set Membership: |
|

LOC4338081
Genomic Map