Assay Details
| Gene Symbol: LOC9269143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784246.2 | XP_015639732.1 | 3 - 4 | 954 | 97 | |
| XM_015784247.2 | XP_015639733.1 | 3 - 4 | 981 | 97 | ||
| XM_015784249.2 | XP_015639735.1 | 4 - 5 | 1128 | 97 | ||
| XM_015784250.2 | XP_015639736.1 | 4 - 5 | 1152 | 97 | ||
More Information
Set Membership: |
|

LOC9269143
Genomic Map