Assay Details
| Gene Symbol: LOC4338349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784812.2 | XP_015640298.1 | 3 - 4 | 352 | 69 | |
| XM_015784813.2 | XP_015640299.1 | 3 - 4 | 205 | 69 | ||
| GenBank mRNA | AK073928.1 | - | 3 - 4 | 357 | 69 | |
More Information
Set Membership: |
|

LOC4338349
Genomic Map