Assay Details
| Gene Symbol: LOC4338349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784812.2 | XP_015640298.1 | 4 - 5 | 522 | 73 | |
| XM_015784813.2 | XP_015640299.1 | 4 - 5 | 375 | 73 | ||
| GenBank mRNA | AK073928.1 | - | 4 - 5 | 527 | 73 | |
More Information
Set Membership: |
|

LOC4338349
Genomic Map