Assay Details
| Gene Symbol: LOC4338384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783408.2 | XP_015638894.1 | 1 | 1958 | 81 | |
| GenBank mRNA | AK107553.1 | - | 1 | 1846 | 81 | |
More Information
Set Membership: |
|

LOC4338384
Genomic Map