Assay Details
| Gene Symbol: LOC4338600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781943.2 | XP_015637429.1 | 3 - 4 | 487 | 140 | |
| GenBank mRNA | AK073553.1 | - | 2 - 3 | 1514 | 140 | |
| AK073796.1 | - | 3 - 4 | 494 | 140 | ||
More Information
Set Membership: |
|

LOC4338600
Genomic Map