Assay Details
| Gene Symbol: LOC4338600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781943.2 | XP_015637429.1 | 7 - 8 | 1214 | 111 | |
| GenBank mRNA | AK073796.1 | - | 7 - 8 | 1221 | 111 | |
More Information
Set Membership: |
|

LOC4338600
Genomic Map