Assay Details
| Gene Symbol: LOC4338790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784691.2 | XP_015640177.1 | 1 - 2 | 123 | 98 | |
| GenBank mRNA | AB104732.1 | - | 1 - 2 | 68 | 98 | |
| AK122099.1 | - | 1 - 2 | 61 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784691.2 transcripts.
Set Membership: |
|

LOC4338790
Genomic Map