Assay Details
| Gene Symbol: LOC4339247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781910.2 | XP_015637396.1 | 10 - 11 | 822 | 74 | |
| GenBank mRNA | AK068335.1 | - | 10 - 11 | 804 | 74 | |
| AK104049.1 | - | 10 - 11 | 802 | 74 | ||
More Information
Set Membership: |
|

LOC4339247
Genomic Map