Assay Details
| Gene Symbol: LOC107276549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783836.2 | XP_015639322.2 | 1 | 873 | 74 | |
| GenBank mRNA | AK100082.1 | - | 1 | 843 | 74 | |
More Information
Set Membership: |
|

LOC107276549
Genomic Map