Assay Details
| Gene Symbol: LOC4339435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784874.2 | XP_015640360.1 | 1 - 2 | 287 | 60 | |
| XM_015784876.2 | XP_015640362.1 | 1 - 2 | 289 | 60 | ||
| GenBank mRNA | AK287614.1 | - | 1 - 2 | 263 | 60 | |
More Information
Set Membership: |
|

LOC4339435
Genomic Map