Assay Details
| Gene Symbol: LOC4339494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784707.2 | XP_015640193.2 | 10 - 11 | 1280 | 94 | |
| GenBank mRNA | AK061952.1 | - | 6 - 7 | 586 | 94 | |
| AK066240.1 | - | 9 - 10 | 1277 | 94 | ||
More Information
Set Membership: |
|

LOC4339494
Genomic Map