Assay Details
| Gene Symbol: LOC4339610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783914.2 | XP_015639400.1 | 16 - 17 | 5072 | 116 | |
| XM_015783915.2 | XP_015639401.1 | 16 - 17 | 5077 | 116 | ||
| GenBank mRNA | AK059222.1 | - | 3 - 4 | 623 | 116 | |
| AK121237.1 | - | 12 - 13 | 3092 | 116 | ||
More Information
Set Membership: |
|

LOC4339610
Genomic Map