Assay Details
| Gene Symbol: LOC4339610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783914.2 | XP_015639400.1 | 17 - 18 | 5333 | 107 | |
| GenBank mRNA | AK059222.1 | - | 4 - 5 | 884 | 107 | |
| AK121237.1 | - | 13 - 14 | 3353 | 107 | ||
More Information
Set Membership: |
|

LOC4339610
Genomic Map