Assay Details
| Gene Symbol: LOC4339610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783914.2 | XP_015639400.1 | 8 - 9 | 2493 | 81 | |
| XM_015783915.2 | XP_015639401.1 | 8 - 9 | 2498 | 81 | ||
| GenBank mRNA | AK121237.1 | - | 4 - 5 | 513 | 81 | |
More Information
Set Membership: |
|

LOC4339610
Genomic Map