Assay Details
| Gene Symbol: LOC4339610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783914.2 | XP_015639400.1 | 10 - 11 | 2714 | 76 | |
| XM_015783915.2 | XP_015639401.1 | 10 - 11 | 2719 | 76 | ||
| GenBank mRNA | AK121237.1 | - | 6 - 7 | 734 | 76 | |
More Information
Set Membership: |
|

LOC4339610
Genomic Map