Assay Details
| Gene Symbol: LOC4339639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784085.2 | XP_015639571.1 | 12 - 13 | 1525 | 81 | |
| GenBank mRNA | AK066453.1 | - | 11 - 12 | 2047 | 81 | |
| AK067742.1 | - | 12 - 13 | 1416 | 81 | ||
| AK098906.1 | - | 12 - 13 | 1413 | 81 | ||
| AK098911.1 | - | 12 - 13 | 1418 | 81 | ||
| AK100749.1 | - | 7 - 8 | 624 | 81 | ||
| AK103421.1 | - | 12 - 13 | 1416 | 81 | ||
| AK104838.1 | - | 12 - 13 | 1415 | 81 | ||
| AK243002.1 | - | 4236 | 81 | |||
| AK289096.1 | - | 4237 | 81 | |||
| EU267973.1 | - | 12 - 13 | 1403 | 81 | ||
More Information
Set Membership: |
|

LOC4339639
Genomic Map