Assay Details
| Gene Symbol: LOC4339639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784085.2 | XP_015639571.1 | 6 - 7 | 972 | 86 | |
| GenBank mRNA | AK066453.1 | - | 5 - 6 | 1494 | 86 | |
| AK067742.1 | - | 6 - 7 | 863 | 86 | ||
| AK103421.1 | - | 6 - 7 | 863 | 86 | ||
| AK104838.1 | - | 6 - 7 | 862 | 86 | ||
| AK243002.1 | - | 3684 | 86 | |||
| AK289096.1 | - | 3684 | 86 | |||
| EU267973.1 | - | 6 - 7 | 850 | 86 | ||
More Information
Set Membership: |
|

LOC4339639
Genomic Map