Assay Details
| Gene Symbol: LOC4339639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784085.2 | XP_015639571.1 | 9 - 10 | 1184 | 101 | |
| GenBank mRNA | AK067742.1 | - | 9 - 10 | 1075 | 101 | |
| AK098906.1 | - | 9 - 10 | 1072 | 101 | ||
| AK098911.1 | - | 9 - 10 | 1077 | 101 | ||
| AK100749.1 | - | 4 - 5 | 283 | 101 | ||
| AK103421.1 | - | 9 - 10 | 1075 | 101 | ||
| AK104838.1 | - | 9 - 10 | 1074 | 101 | ||
| AK243002.1 | - | 3896 | 101 | |||
| AK289096.1 | - | 3896 | 101 | |||
| EU267973.1 | - | 9 - 10 | 1062 | 101 | ||
More Information
Set Membership: |
|

LOC4339639
Genomic Map