Assay Details
| Gene Symbol: LOC4339846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787545.2 | XP_015643031.1 | 10 - 11 | 1740 | 78 | |
| GenBank mRNA | AK059993.1 | - | 2 - 3 | 176 | 78 | |
| AK060849.1 | - | 10 - 11 | 1599 | 78 | ||
| AK098983.1 | - | 10 - 11 | 1539 | 78 | ||
| AK100758.1 | - | 10 - 11 | 1596 | 78 | ||
| AK104349.1 | - | 2 - 3 | 176 | 78 | ||
More Information
Set Membership: |
|

LOC4339846
Genomic Map