Assay Details
| Gene Symbol: LOC4339846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787545.2 | XP_015643031.1 | 11 - 12 | 1854 | 84 | |
| GenBank mRNA | AK059993.1 | - | 3 - 4 | 290 | 84 | |
| AK060849.1 | - | 11 - 12 | 1713 | 84 | ||
| AK098983.1 | - | 11 - 12 | 1653 | 84 | ||
| AK100758.1 | - | 11 - 12 | 1710 | 84 | ||
| AK104349.1 | - | 3 - 4 | 290 | 84 | ||
More Information
Set Membership: |
|

LOC4339846
Genomic Map