Assay Details
| Gene Symbol: LOC4339846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787545.2 | XP_015643031.1 | 12 - 13 | 1926 | 112 | |
| GenBank mRNA | AK059993.1 | - | 4 - 5 | 362 | 112 | |
| AK060849.1 | - | 12 - 13 | 1785 | 112 | ||
| AK098983.1 | - | 12 - 13 | 1725 | 112 | ||
| AK100758.1 | - | 12 - 13 | 1782 | 112 | ||
| AK104349.1 | - | 4 - 5 | 362 | 112 | ||
More Information
Set Membership: |
|

LOC4339846
Genomic Map