Assay Details
| Gene Symbol: LOC4339846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787545.2 | XP_015643031.1 | 3 - 4 | 628 | 112 | |
| GenBank mRNA | AK060849.1 | - | 3 - 4 | 487 | 112 | |
| AK098983.1 | - | 3 - 4 | 427 | 112 | ||
| AK100758.1 | - | 3 - 4 | 484 | 112 | ||
More Information
Set Membership: |
|

LOC4339846
Genomic Map