Assay Details
| Gene Symbol: LOC4339846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787545.2 | XP_015643031.1 | 7 - 8 | 1297 | 99 | |
| GenBank mRNA | AK098983.1 | - | 7 - 8 | 1096 | 99 | |
| AK100758.1 | - | 7 - 8 | 1153 | 99 | ||
More Information
Set Membership: |
|

LOC4339846
Genomic Map