Assay Details
| Gene Symbol: LOC4339846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787545.2 | XP_015643031.1 | 9 - 10 | 1624 | 95 | |
| GenBank mRNA | AK059993.1 | - | 1 - 2 | 60 | 95 | |
| AK098983.1 | - | 9 - 10 | 1423 | 95 | ||
| AK100758.1 | - | 9 - 10 | 1480 | 95 | ||
| AK104349.1 | - | 1 - 2 | 60 | 95 | ||
More Information
Set Membership: |
|

LOC4339846
Genomic Map